Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038997.2 | NP_001034086.1 | 2, 3 | 230 | ||
GenBank mRNA | BC132026.1 | - | 2, 3 | 230 | 40130303 | |
BC132030.1 | - | 2, 3 | 230 | 40130307 |
Gene Symbol: Gm10334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103153.1 | NP_001096623.1 | 2, 3 | 232 |
Gene Symbol: Prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011645.2 | NP_035775.1 | 2, 3 | 216 | ||
GenBank mRNA | AK131862.1 | - | 2, 3 | 217 | ||
BC098498.1 | - | 2, 3 | 199 | |||
BC141304.1 | - | 2, 3 | 281 | 9055916 |
Gene Symbol: Try5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003405.4 | NP_001003405.1 | 2, 3 | 215 | ||
GenBank mRNA | AB009661.1 | - | 2, 3 | 215 | ||
AK007843.1 | - | 2, 3 | 214 | |||
AK131898.1 | - | 2, 3 | 214 | |||
AK160484.1 | - | 2, 3 | 214 | |||
BC139220.1 | - | 2, 3 | 300 | 8862242 | ||
BC139223.1 | - | 2, 3 | 300 | 8862245 | ||
BY061854.1 | - | 2, 3 | 224 |