Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Try4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011646.5 | NP_035776.1 | 3 | 303 | ||
GenBank mRNA | AK003064.1 | - | 3 | 307 | ||
AK008660.1 | - | 3 | 304 | |||
AK008667.1 | - | 3 | 304 | |||
BC050120.1 | - | 3 | 284 | |||
BC061135.1 | - | 3 | 303 | 30306436 |
Gene Symbol: Try5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003405.4 | NP_001003405.1 | 3 | 305 | ||
GenBank mRNA | AB009661.1 | - | 3 | 305 | ||
AK007843.1 | - | 3 | 304 | |||
AK131898.1 | - | 3 | 304 | |||
AK160484.1 | - | 3 | 304 | |||
BC139220.1 | - | 3 | 390 | 8862242 | ||
BC139223.1 | - | 3 | 390 | 8862245 |