Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039363.2 | NP_001034452.1 | 3 | 297 | ||
NM_001286713.1 | NP_001273642.1 | 3 | 297 | |||
NM_001286714.1 | NP_001273643.1 | 3 | 292 | |||
NM_001286716.1 | NP_001273645.1 | 3 | 292 | |||
NM_001286718.1 | NP_001273647.1 | 3 | 297 | |||
NM_001286720.1 | NP_001273649.1 | 3 | 297 | |||
NM_011647.3 | NP_035777.2 | 3 | 297 | |||
GenBank mRNA | AK143799.1 | - | 3 | 298 | ||
AK147016.1 | - | 3 | 273 | |||
AK147302.1 | - | 3 | 237 | |||
AK147487.1 | - | 3 | 266 | |||
AK147874.1 | - | 3 | 262 | |||
AK147908.1 | - | 3 | 298 | |||
AK148062.1 | - | 3 | 289 | |||
AK148072.1 | - | 3 | 194 | |||
AK170589.1 | - | 3 | 288 | |||
AK171007.1 | - | 3 | 269 | |||
BC052449.1 | - | 3 | 273 | 6409037 | ||
BC060701.1 | - | 3 | 273 | 6409037 | ||
BY742714.1 | - | 3 | 301 | |||
U37775.1 | - | 2 | 212 | |||
U39818.1 | - | 3 | 268 |