Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025290.3 | NP_079566.1 | 8 | 946 | ||
GenBank mRNA | AK005739.1 | - | 8 | 898 | ||
BC049584.1 | - | 8 | 946 | 6744132 | ||
DQ141549.1 | - | 8 | 845 | |||
DQ141550.1 | - | 8 | 845 | |||
DQ141551.1 | - | 8 | 845 | |||
DQ141553.1 | - | 8 | 845 | |||
DQ141554.1 | - | 8 | 845 | |||
DQ141555.1 | - | 7 | 731 | |||
DQ141556.1 | - | 7 | 731 |