Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002207.2 | NP_002198.2 | 8 | 921 | ||
GenBank mRNA | BC030198.1 | - | 8 | 1128 | 5199296 | |
D25303.1 | - | 9 | 919 | |||
L24158.1 | - | 8 | 763 |