Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009442.2 | NP_033468.2 | 11 | 2573 | ||
GenBank mRNA | AF237703.1 | - | 10 | 2525 | ||
AK140081.1 | - | 10 | 3791 | |||
BC059011.1 | - | 10 | 2540 | 6847867 | ||
X83974.1 | - | 12 | 2481 |