Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110265.1 | NP_001103735.1 | 19 | 2263 | ||
NM_001284272.1 | NP_001271201.1 | 18 | 2092 | |||
NM_009445.2 | NP_033471.2 | 19 | 2263 | |||
GenBank mRNA | AK041487.1 | - | 19 | 2234 | ||
AK164093.1 | - | 18 | 2091 | |||
AX775731.1 | - | 19 | 2229 | |||
BC058851.1 | - | 19 | 2249 | 30023533 | ||
DT914108.1 | - | 3 | 224 | |||
M86377.1 | - | 20 | 2289 |