Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009450.2 | NP_033476.1 | 4 | 434 | ||
GenBank mRNA | AK134545.1 | - | 4 | 433 | ||
BC055441.1 | - | 4 | 424 | 4016867 |