Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011655.5 | NP_035785.1 | 4 | 1500 | ||
GenBank mRNA | AK010960.1 | - | 4 | 1432 | ||
AK011263.1 | - | 4 | 1428 | |||
AK011765.1 | - | 4 | 1429 | |||
AK012350.1 | - | 4 | 1429 | |||
AK012365.1 | - | 2 | 1070 | |||
AK013283.1 | - | 2 | 1060 | |||
AK017806.1 | - | 4 | 1429 | |||
AK051164.1 | - | 4 | 1432 | |||
AK051393.1 | - | 4 | 1434 | |||
AK076196.1 | - | 4 | 1413 | |||
AK083327.1 | - | 4 | 1432 | |||
AK146567.1 | - | 4 | 1432 | |||
AK148978.1 | - | 4 | 1427 | |||
AK151215.1 | - | 4 | 1432 | |||
AK151670.1 | - | 4 | 1432 | |||
AK160225.1 | - | 4 | 1432 | |||
AK161575.1 | - | 4 | 1432 | |||
AK165249.1 | - | 4 | 1430 | |||
AK167691.1 | - | 4 | 1432 | |||
AK167779.1 | - | 4 | 1432 | |||
AK167808.1 | - | 4 | 1427 | |||
AK167926.1 | - | 4 | 1432 | |||
AK168271.1 | - | 4 | 1427 | |||
AK169123.1 | - | 4 | 1427 | |||
AK169210.1 | - | 4 | 1431 | |||
AK169295.1 | - | 4 | 1432 | |||
AK169398.1 | - | 1 | 909 | |||
AK169606.1 | - | 4 | 1431 | |||
BC003825.1 | - | 4 | 1406 | 3601177 | ||
BC047993.1 | - | 1 | 480 |