Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293728.1 | NP_001280657.1 | 4 | 375 | ||
NM_011656.3 | NP_035786.1 | 5 | 450 | |||
GenBank mRNA | AK132488.1 | - | 5 | 382 |