Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twist1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011658.2 | NP_035788.1 | 2 | 1028 | ||
GenBank mRNA | BC033434.1 | - | 2 | 854 | 4935230 | |
BC083139.1 | - | 2 | 1028 | 6516673 | ||
M63650.1 | - | 2 | 887 |