Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011659.2 | NP_035789.1 | 3 | 534 | ||
GenBank mRNA | BC139239.1 | - | 3 | 404 | 8862261 | |
BC139266.1 | - | 3 | 404 | 8862288 | ||
Z21674.1 | - | 3 | 355 |