Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122754.2 | NP_001116226.1 | 10 | 919 | ||
NM_001289494.1 | NP_001276423.1 | 9 | 853 | |||
NM_001289495.1 | NP_001276424.1 | 10 | 936 | |||
NM_013698.3 | NP_038726.2 | 9 | 1003 | |||
GenBank mRNA | AK088209.1 | - | 9 | 849 | ||
AK138069.1 | - | 10 | 916 | |||
AK138786.1 | - | 10 | 933 | |||
BC120552.1 | - | 9 | 906 | 40129475 | ||
BC137710.1 | - | 9 | 908 | 8860729 | ||
D43963.1 | - | 9 | 1003 | |||
D83201.1 | - | 9 | 943 | |||
D83202.1 | - | 9 | 1106 | |||
U16145.1 | - | 10 | 884 | |||
U19607.1 | - | 10 | 919 |