Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011660.3 | NP_035790.1 | 3 | 358 | ||
GenBank mRNA | AK007537.1 | - | 3 | 239 | ||
AK007790.1 | - | 3 | 236 | |||
BC010756.1 | - | 3 | 213 | 4216106 | ||
BC094415.1 | - | 3 | 220 | 3376881 | ||
X77585.1 | - | 3 | 231 |