Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011661.4 | NP_035791.1 | 2 | 949 | ||
GenBank mRNA | AK014619.1 | - | 2 | 989 | ||
AK148172.1 | - | 2 | 972 | |||
AK148341.1 | - | 2 | 899 | |||
AK148357.1 | - | 2 | 972 | |||
AK148432.1 | - | 2 | 928 | |||
AY526904.1 | - | 2 | 990 | |||
BC079678.1 | - | 2 | 899 | 30539846 | ||
D00131.1 | - | 2 | 952 | |||
D00440.1 | - | 2 | 949 | |||
M20234.1 | - | 2 | 1050 | |||
M24560.1 | - | 2 | 949 | |||
M26729.1 | - | 2 | 888 | |||
X12782.1 | - | 2 | 990 |