Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290800.1 | NP_001277729.1 | 14 | 2394 | ||
NM_019392.2 | NP_062265.2 | 14 | 1930 | |||
GenBank mRNA | AB000828.1 | - | 14 | 1667 | ||
AK134607.1 | - | 6 | 767 | |||
AK141198.1 | - | 14 | 1701 | |||
BC066058.1 | - | 14 | 1930 | 6834015 | ||
BC082325.1 | - | 14 | 1878 | 30356080 | ||
D17393.1 | - | 14 | 2372 | |||
U05683.1 | - | 14 | 1715 | |||
U18342.1 | - | 14 | 1847 | |||
U18343.1 | - | 14 | 2390 | |||
U18933.1 | - | 14 | 1866 | |||
X78103.1 | - | 14 | 1874 |