Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282014.1 | NP_001268943.1 | 8 | 1695 | ||
NM_001282015.1 | NP_001268944.1 | 8 | 1689 | |||
NM_031202.3 | NP_112479.1 | 8 | 1710 | |||
GenBank mRNA | AK148332.1 | - | 8 | 1695 | ||
AK148370.1 | - | 8 | 1601 | |||
AK148441.1 | - | 8 | 1601 | |||
AK181989.1 | - | 3 | 194 | |||
AK188091.1 | - | 3 | 196 | |||
AK189039.1 | - | 3 | 197 | |||
AK190379.1 | - | 2 | 176 | |||
AK192902.1 | - | 2 | 171 | |||
AK193585.1 | - | 3 | 196 | |||
AK194427.1 | - | 3 | 196 | |||
AK194794.1 | - | 2 | 173 | |||
AK194981.1 | - | 2 | 176 | |||
AK196132.1 | - | 2 | 173 | |||
AK197740.1 | - | 3 | 196 | |||
AK198835.1 | - | 3 | 196 | |||
AK198900.1 | - | 2 | 178 | |||
AK199149.1 | - | 3 | 196 | |||
AK199163.1 | - | 3 | 196 | |||
AK199776.1 | - | 3 | 196 | |||
AK199872.1 | - | 3 | 196 | |||
AK200013.1 | - | 3 | 196 | |||
AK200321.1 | - | 3 | 180 | |||
AK200365.1 | - | 3 | 196 | |||
AK200395.1 | - | 3 | 196 | |||
AK202380.1 | - | 3 | 189 | |||
AK203562.1 | - | 2 | 151 | |||
AK204415.1 | - | 2 | 164 | |||
AK207921.1 | - | 3 | 209 | |||
BC076598.1 | - | 8 | 1693 | 30613975 | ||
X03687.1 | - | 8 | 1595 |