Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136085.2 | NP_001129557.1 | 24 | 3103 | ||
NM_001276316.1 | NP_001263245.1 | 25 | 3195 | |||
NM_001276317.1 | NP_001263246.1 | 24 | 3098 | |||
NM_009457.4 | NP_033483.2 | 24 | 3098 | |||
GenBank mRNA | AK088057.1 | - | 24 | 2980 | ||
AK088528.1 | - | 24 | 2980 | |||
AK143416.1 | - | 24 | 3004 | |||
AK171667.1 | - | 24 | 3069 | |||
AK182461.1 | - | 2 | 229 | |||
AK188980.1 | - | 2 | 202 | |||
AK199544.1 | - | 4 | 354 | |||
AK202478.1 | - | 4 | 354 | |||
AK204860.1 | - | 4 | 354 | |||
AK205991.1 | - | 2 | 187 | |||
AK211490.1 | - | 2 | 189 | |||
AK211788.1 | - | 2 | 137 | |||
BC058630.1 | - | 24 | 3044 | 6402368 | ||
BC138200.1 | - | 24 | 2961 | 8861219 | ||
BC145984.1 | - | 24 | 2962 | 40131287 | ||
D10576.1 | - | 24 | 2964 | |||
X62580.1 | - | 9 | 1033 |