Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009458.4 | NP_033484.3 | 4 | 345 | ||
GenBank mRNA | AK010432.1 | - | 4 | 243 | ||
AK011363.1 | - | 4 | 266 | |||
AK147785.1 | - | 4 | 346 | |||
AK152316.1 | - | 4 | 328 | |||
AK152645.1 | - | 4 | 328 | |||
AK153174.1 | - | 4 | 328 | |||
AK169229.1 | - | 4 | 328 | |||
BC132075.1 | - | 4 | 266 | 40130352 | ||
BC132077.1 | - | 4 | 266 | 40130354 | ||
U57690.1 | - | 4 | 258 | |||
X96859.1 | - | 4 | 242 |