Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019803.3 | NP_062777.2 | 3 | 167 | ||
GenBank mRNA | AF296657.1 | - | 3 | 148 | ||
AK136365.1 | - | 3 | 154 | |||
AK146554.1 | - | 3 | 172 | |||
AK154464.1 | - | 3 | 188 | |||
AK183367.1 | - | 3 | 120 | |||
AK185288.1 | - | 3 | 110 | |||
BC010321.1 | - | 3 | 167 | 2651046 | ||
U93241.1 | - | 2 | 67 |