Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009460.2 | NP_033486.1 | 4, 5 | 381 | ||
GenBank mRNA | AF033353.1 | - | 4, 5 | 356 | ||
AK002536.1 | - | 4, 5 | 351 | |||
AK011074.1 | - | 4, 5 | 350 | |||
AK089081.1 | - | 4, 5 | 330 | |||
AK159366.1 | - | 4, 5 | 352 | |||
AK220052.1 | - | 3, 4 | 245 | |||
BC082566.1 | - | 4, 5 | 356 | 30602751 | ||
BC083158.1 | - | 4, 5 | 354 | 6478032 |