Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009466.2 | NP_033492.1 | 8 | 1177 | ||
GenBank mRNA | AF061017.1 | - | 8 | 1061 | ||
AK144673.1 | - | 8 | 1213 | |||
AK146693.1 | - | 8 | 1061 | |||
AK149590.1 | - | 2 | 2457 | |||
AK162298.1 | - | 8 | 1065 | |||
AK167462.1 | - | 8 | 1057 | |||
AK167560.1 | - | 8 | 1157 | |||
AK168064.1 | - | 6 | 774 | |||
AK168508.1 | - | 8 | 1064 | |||
AK168563.1 | - | 8 | 1061 | |||
AK169208.1 | - | 8 | 1192 | |||
BC006749.1 | - | 8 | 1177 | 3708709 |