Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009467.3 | NP_033493.3 | 2 | 776 | ||
GenBank mRNA | AK136480.1 | - | 2 | 777 | ||
BC028262.1 | - | 2 | 767 | 5102040 | ||
X06358.1 | - | 2 | 754 |