Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011674.4 | NP_035804.2 | 6 | 1641 | ||
GenBank mRNA | AK137364.1 | - | 6 | 1642 | ||
AK144005.1 | - | 6 | 1641 | |||
AK163650.1 | - | 6 | 1603 | |||
BC016885.1 | - | 1636 | 4223057 |