Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011676.2 | NP_035806.1 | 2 | 293 | ||
GenBank mRNA | AF030169.1 | - | 2 | 317 | ||
AK131955.1 | - | 2 | 298 | |||
BC001990.1 | - | 2 | 293 | 3484396 |