Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011678.2 | NP_035808.2 | 20 | 2676 | ||
GenBank mRNA | AK143582.1 | - | 20 | 2611 | ||
AK149964.1 | - | 20 | 2634 | |||
AK163201.1 | - | 3 | 322 | |||
AK164245.1 | - | 11 | 1451 | |||
AK169372.1 | - | 21 | 2655 | |||
AK169933.1 | - | 20 | 2646 | |||
AK171271.1 | - | 20 | 2646 | |||
AK171633.1 | - | 20 | 2629 | |||
AK216305.1 | - | 2 | 90 | |||
BC011341.1 | - | 19 | 2453 | 3661165 | ||
BC066180.1 | - | 20 | 2586 | |||
BC066865.1 | - | 20 | 2586 | |||
L00681.1 | - | 20 | 2639 |