Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011678.2 | NP_035808.2 | 9 | 1206 | ||
GenBank mRNA | AK143582.1 | - | 9 | 1141 | ||
AK149964.1 | - | 9 | 1164 | |||
AK169372.1 | - | 9 | 1185 | |||
AK169933.1 | - | 9 | 1176 | |||
AK171271.1 | - | 9 | 1176 | |||
AK171633.1 | - | 9 | 1159 | |||
AK220512.1 | - | 9 | 1163 | |||
BC011341.1 | - | 8 | 983 | 3661165 | ||
BC066180.1 | - | 9 | 1116 | |||
BC066865.1 | - | 9 | 1116 | |||
L00681.1 | - | 9 | 1169 |