Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009476.2 | NP_033502.2 | 3, 4 | 400 | ||
GenBank mRNA | BC104260.1 | - | 3, 4 | 400 | 40039813 | |
BC104261.1 | - | 3, 4 | 400 | 40039814 | ||
DQ358978.1 | - | 3, 4 | 339 | |||
U08030.1 | - | 3, 4 | 398 |