Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000212.2 | NP_000203.2 | 11 | 1769 | ||
GenBank mRNA | BC127666.1 | - | 11 | 1769 | 40128462 | |
BC127667.1 | - | 11 | 1773 | 40128464 | ||
J02703.1 | - | 11 | 1769 | |||
KC120777.1 | - | 11 | 1770 | |||
KC120778.1 | - | 11 | 1769 | |||
M20311.1 | - | 11 | 1765 | |||
M35999.1 | - | 11 | 1767 | |||
U95204.1 | - | 11 | 1749 |