Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009478.3 | NP_033504.2 | 6 | 625 | ||
GenBank mRNA | AK002844.1 | - | 6 | 615 | ||
AK007470.1 | - | 6 | 615 | |||
AK135090.1 | - | 2 | 457 | |||
AK172426.1 | - | 6 | 636 | |||
AK211828.1 | - | 1 | 128 | |||
BC008109.1 | - | 6 | 625 | 3481699 |