Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011680.2 | NP_035810.1 | 3 | 233 | ||
GenBank mRNA | AB221562.1 | - | 3 | 192 | ||
AK146871.1 | - | 3 | 180 | |||
BC082995.1 | - | 3 | 233 | 6440967 | ||
U01662.1 | - | 3 | 249 |