Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ush2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021408.3 | NP_067383.3 | 39 | 7658 | ||
GenBank mRNA | DQ073638.1 | - | 39 | 7462 |