Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000213.4 | NP_000204.3 | 30, 31 | 4080 | ||
NM_001005619.1 | NP_001005619.1 | 29, 30 | 3793 | |||
NM_001005731.2 | NP_001005731.1 | 30, 31 | 4080 | |||
NM_001321123.1 | NP_001308052.1 | 30, 31 | 4031 | |||
XM_005257309.2 | XP_005257366.1 | 30, 31 | 3850 | |||
XM_005257311.4 | XP_005257368.1 | 30, 31 | 4069 | |||
XM_006721866.3 | XP_006721929.1 | 30, 31 | 3996 | |||
XM_006721867.3 | XP_006721930.1 | 30, 31 | 3993 | |||
XM_006721868.3 | XP_006721931.1 | 30, 31 | 3990 | |||
XM_006721870.3 | XP_006721933.1 | 30, 31 | 3986 | |||
XM_011524751.2 | XP_011523053.1 | 30, 31 | 3994 | |||
XM_011524752.2 | XP_011523054.1 | 15, 16 | 2279 | |||
GenBank mRNA | AB208913.1 | - | 30, 31 | 3882 | ||
BC118916.1 | - | 30, 31 | 3842 | 40073279 | ||
BC126411.1 | - | 30, 31 | 3901 | 8992127 | ||
BC143738.1 | - | 30, 31 | 3901 | 9052252 | ||
BC143742.1 | - | 30, 31 | 3852 | 9052256 | ||
X51841.1 | - | 30, 31 | 3911 | |||
X52186.1 | - | 29, 30 | 3794 | |||
X53587.1 | - | 30, 31 | 3972 |