Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011682.4 | NP_035812.3 | 16 | 2047 | ||
GenBank mRNA | AK134010.1 | - | 16 | 2041 | ||
AK134424.1 | - | 17 | 2659 | |||
AK163051.1 | - | 16 | 2050 | |||
BC011062.1 | - | 17 | 2096 | |||
BC024140.1 | - | 8 | 1081 | |||
BC050183.1 | - | 17 | 2156 | |||
BC062163.1 | - | 17 | 2103 | |||
Y12229.1 | - | 16 | 1958 |