Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009489.2 | NP_033515.2 | 1 | 241 | ||
GenBank mRNA | AF011424.1 | - | 1 | 241 |
Gene Symbol: Vmn2r32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105063.1 | NP_001098533.1 | 1 | 126 |
Gene Symbol: Vmn2r34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105066.1 | NP_001098536.1 | 1 | 126 |
Gene Symbol: Vmn2r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009493.2 | NP_033519.1 | 1 | 242 | ||
GenBank mRNA | AF011414.1 | - | 1 | 242 |
Gene Symbol: Vmn2r45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105075.1 | NP_001098545.1 | 1 | 126 |
Gene Symbol: Vmn2r51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105179.1 | NP_001098649.1 | 1 | 126 |
Set Membership: |
G-protein coupled receptor |