Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009489.2 | NP_033515.2 | 1 | 203 | ||
GenBank mRNA | AF011424.1 | - | 1 | 203 |
Gene Symbol: Vmn2r30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009490.3 | NP_033516.3 | 1 | 88 | ||
GenBank mRNA | AF011425.1 | - | 126 |
Gene Symbol: Vmn2r32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105063.1 | NP_001098533.1 | 1 | 88 |
Gene Symbol: Vmn2r34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105066.1 | NP_001098536.1 | 1 | 88 |
Gene Symbol: Vmn2r36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105068.1 | NP_001098538.1 | 1 | 88 |
Gene Symbol: Vmn2r40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105072.1 | NP_001098542.1 | 1 | 88 |
Gene Symbol: Vmn2r41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105073.1 | NP_001098543.1 | 1 | 88 |
Gene Symbol: Vmn2r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009493.2 | NP_033519.1 | 1 | 204 | ||
GenBank mRNA | AF011414.1 | - | 1 | 204 |
Gene Symbol: Vmn2r45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105075.1 | NP_001098545.1 | 1 | 88 |
Gene Symbol: Vmn2r47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105151.1 | NP_001098621.1 | 1 | 88 |
Gene Symbol: Vmn2r48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105152.1 | NP_001098622.1 | 1 | 97 |
Gene Symbol: Vmn2r49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105156.1 | NP_001098626.1 | 1 | 88 |
Gene Symbol: Vmn2r51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105179.1 | NP_001098649.1 | 1 | 88 |
Set Membership: |
G-protein coupled receptor |