Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313941.1 | NP_001300870.1 | 6 | 1115 | ||
NM_001313943.1 | NP_001300872.1 | 6 | 1159 | |||
NM_001313944.1 | NP_001300873.1 | 5 | 960 | |||
NM_001313945.1 | NP_001300874.1 | 5 | 897 | |||
NM_001313946.1 | NP_001300875.1 | 3 | 768 | |||
NM_001313947.1 | NP_001300876.1 | 4 | 899 | |||
NM_001664.3 | NP_001655.1 | 5 | 1020 | |||
GenBank mRNA | AK130066.1 | - | 5 | 1016 | ||
AK130808.1 | - | 5 | 896 | |||
AK222556.1 | - | 5 | 912 | |||
AK296654.1 | - | 5 | 836 | |||
AK307220.1 | - | 5 | 795 | |||
BC000946.3 | - | 2 | 464 | |||
BC001360.2 | - | 5 | 852 | 3051460 | ||
BC005976.1 | - | 5 | 857 | 4102976 | ||
BU509012.1 | - | 4 | 730 | |||
BX647063.1 | - | 5 | 916 | |||
L09159.1 | - | 4 | 729 | |||
L25080.1 | - | 5 | 891 | |||
X05026.1 | - | 5 | 898 |