Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009502.4 | NP_033528.3 | 21 | 3155 | ||
GenBank mRNA | AB041262.1 | - | 3 | 176 | ||
AK035184.1 | - | 8 | 1186 | |||
AK077850.1 | - | 21 | 3191 | |||
BC008520.1 | - | 21 | 3155 | 3495710 | ||
BC008554.1 | - | 7 | 1036 | |||
L18880.1 | - | 21 | 3084 |