Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009502.4 | NP_033528.3 | 18 | 2821 | ||
GenBank mRNA | AK035184.1 | - | 5 | 852 | ||
AK077850.1 | - | 18 | 2857 | |||
BC008520.1 | - | 18 | 2821 | 3495710 | ||
BC008554.1 | - | 4 | 702 | |||
L18880.1 | - | 18 | 2750 |