Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000888.4 | NP_000879.2 | 6 | 1109 | ||
NM_001282353.1 | NP_001269282.1 | 7 | 1002 | |||
NM_001282354.1 | NP_001269283.1 | 4 | 824 | |||
NM_001282355.1 | NP_001269284.1 | 6 | 1109 | |||
NM_001282388.1 | NP_001269317.1 | 5 | 1029 | |||
NM_001282389.1 | NP_001269318.1 | 6 | 936 | |||
NM_001282390.1 | NP_001269319.1 | 5 | 856 | |||
GenBank mRNA | AK290300.1 | - | 6 | 1077 | ||
AK304203.1 | - | 5 | 968 | |||
AK313944.1 | - | 7 | 993 | |||
AL831998.1 | - | 6 | 1048 | |||
BC121178.2 | - | 6 | 879 | 40122759 | ||
BC143656.1 | - | 6 | 887 | |||
CD013944.1 | - | 5 | 801 | |||
CD013945.1 | - | 6 | 1054 | |||
CD013946.1 | - | 4 | 769 | |||
CD013947.1 | - | 6 | 881 | |||
M35198.3 | - | 6 | 875 |