Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011695.2 | NP_035825.1 | 9 | 906 | ||
GenBank mRNA | AK012359.1 | - | 9 | 904 | ||
AK078555.1 | - | 9 | 911 | |||
AK150504.1 | - | 9 | 932 | |||
AK150627.1 | - | 9 | 932 | |||
AK150940.1 | - | 9 | 924 | |||
AK152274.1 | - | 9 | 904 | |||
AK159561.1 | - | 9 | 1141 | |||
AK168199.1 | - | 9 | 877 | |||
AK168328.1 | - | 9 | 903 | |||
BC049359.1 | - | 1 | 1644 | |||
U30838.1 | - | 9 | 906 |