Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000888.4 | NP_000879.2 | 11 | 2089 | ||
NM_001282353.1 | NP_001269282.1 | 12 | 1982 | |||
NM_001282354.1 | NP_001269283.1 | 9 | 1804 | |||
NM_001282388.1 | NP_001269317.1 | 10 | 2009 | |||
NM_001282389.1 | NP_001269318.1 | 11 | 1916 | |||
NM_001282390.1 | NP_001269319.1 | 10 | 1836 | |||
GenBank mRNA | AK290300.1 | - | 11 | 2057 | ||
AK304203.1 | - | 10 | 1948 | |||
AK313944.1 | - | 12 | 1973 | |||
AL831998.1 | - | 11 | 2028 | |||
BC121178.2 | - | 11 | 1859 | 40122759 | ||
BC143656.1 | - | 8 | 1224 | |||
CD013944.1 | - | 10 | 1781 | |||
CD013946.1 | - | 9 | 1749 | |||
CD013947.1 | - | 11 | 1861 | |||
M35198.3 | - | 11 | 1855 |