Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009506.2 | NP_033532.1 | 4 | 897 | ||
GenBank mRNA | AK047844.1 | - | 824 | |||
BC096377.1 | - | 4 | 802 | 5002720 | ||
U58112.1 | - | 4 | 793 | |||
U73620.1 | - | 4 | 803 |