Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011698.1 | NP_035828.1 | 4 | 289 | ||
GenBank mRNA | AF087694.1 | - | 4 | 296 | ||
AF173082.1 | - | 4 | 289 | |||
AK019299.1 | - | 4 | 294 | |||
BC031780.1 | - | 4 | 308 | 4164883 |