Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ezr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009510.2 | NP_033536.2 | 6 | 815 | ||
GenBank mRNA | AK002766.1 | - | 6 | 789 | ||
AK004588.2 | - | 6 | 790 | |||
AK035271.1 | - | 2 | 183 | |||
AK135194.1 | - | 6 | 791 | |||
AK135353.1 | - | 6 | 791 | |||
AK145707.1 | - | 6 | 816 | |||
AK159717.1 | - | 6 | 793 | |||
AK159824.1 | - | 6 | 791 | |||
AK159937.1 | - | 6 | 792 | |||
AK166191.1 | - | 6 | 791 | |||
AK168410.1 | - | 6 | 791 | |||
AK168840.1 | - | 6 | 791 | |||
AK170610.1 | - | 6 | 791 | |||
AK172336.1 | - | 6 | 798 | |||
AK189604.1 | - | 2 | 148 | |||
BC048181.1 | - | 6 | 778 | 6826190 | ||
BC098502.1 | - | 6 | 767 | 30047137 | ||
X60671.1 | - | 6 | 773 |