Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011701.4 | NP_035831.2 | 8 | 1428 | ||
GenBank mRNA | AK012469.1 | - | 8 | 931 | ||
AK076001.1 | - | 8 | 1425 | |||
AK131618.1 | - | 8 | 1430 | |||
AK133726.1 | - | 8 | 1429 | |||
AK150270.1 | - | 8 | 1124 | |||
AK151196.1 | - | 8 | 1431 | |||
AK153019.1 | - | 8 | 1431 | |||
AK159539.1 | - | 8 | 1429 | |||
AK169181.1 | - | 8 | 1430 | |||
AK170429.1 | - | 8 | 1429 | |||
AK203726.1 | - | 4 | 350 | |||
AK213831.1 | - | 4 | 356 | |||
BC089335.1 | - | 8 | 1424 | 6415629 | ||
M24849.1 | - | 8 | 1345 | |||
M26251.1 | - | 8 | 1787 | |||
X51438.1 | - | 8 | 1397 | |||
X56397.1 | - | 8 | 1391 |