Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011709.5 | NP_035839.3 | 2, 3 | 254 | ||
GenBank mRNA | AK142655.1 | - | 255 | |||
AK164735.1 | - | 2, 3 | 255 | |||
BC132280.1 | - | 2, 3 | 252 | 40130557 | ||
BC132282.1 | - | 2, 3 | 252 | 40130559 |