Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164314.1 | NP_001157786.1 | 4, 5 | 605 | ||
NM_001164488.1 | NP_001157960.1 | 4, 5 | 538 | |||
NM_011710.3 | NP_035840.3 | 4, 5 | 551 | |||
GenBank mRNA | AK004541.1 | - | 4, 5 | 539 | ||
AK007754.1 | - | 2, 3 | 167 | |||
AK150090.1 | - | 4, 5 | 569 | |||
AK150267.1 | - | 5, 6 | 653 | |||
AK152966.1 | - | 4, 5 | 569 | |||
AK153337.1 | - | 4, 5 | 576 | |||
AK179345.1 | - | 1, 2 | 26 | |||
AK179848.1 | - | 1, 2 | 26 | |||
AK183767.1 | - | 1, 2 | 26 | |||
AK185391.1 | - | 1, 2 | 26 | |||
AK190023.1 | - | 1, 2 | 26 | |||
AK190625.1 | - | 1, 2 | 26 | |||
AK192489.1 | - | 1, 2 | 26 | |||
AK200381.1 | - | 1, 2 | 26 | |||
AK201963.1 | - | 1, 2 | 26 | |||
AK203296.1 | - | 1, 2 | 26 | |||
BC003450.1 | - | 4, 5 | 537 | 2654231 | ||
BC046232.1 | - | 4, 5 | 544 | 6813406 | ||
CF731228.1 | - | 1, 2 | 57 | |||
CT010258.1 | - | 3, 4 | 437 |