Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016852.2 | NP_058548.1 | 3 | 314 | ||
GenBank mRNA | AK004824.1 | - | 3 | 316 | ||
AK043037.1 | - | 3 | 288 | |||
AK146572.1 | - | 3 | 316 | |||
AK150018.1 | - | 3 | 289 | |||
AK151368.1 | - | 3 | 306 | |||
AK162810.1 | - | 3 | 289 | |||
AK170317.1 | - | 3 | 315 | |||
AK170474.1 | - | 3 | 317 | |||
BC031908.1 | - | 3 | 277 | 5365118 | ||
BC055058.1 | - | 3 | 334 | 6529458 | ||
U40826.1 | - | 3 | 314 |