Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016852.2 | NP_058548.1 | 2 | 132 | ||
GenBank mRNA | AK004824.1 | - | 2 | 134 | ||
AK043037.1 | - | 2 | 106 | |||
AK079300.1 | - | 2 | 122 | |||
AK146572.1 | - | 2 | 134 | |||
AK150018.1 | - | 2 | 107 | |||
AK151368.1 | - | 2 | 124 | |||
AK162810.1 | - | 2 | 107 | |||
AK170317.1 | - | 2 | 133 | |||
AK170474.1 | - | 2 | 135 | |||
BC031908.1 | - | 2 | 95 | 5365118 | ||
BC055058.1 | - | 2 | 152 | 6529458 | ||
U40826.1 | - | 2 | 132 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK044015.1 | - | 122 | Chr. - - [-] on Build |